missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagniSort™ Mouse CD45.2 Positive Selection Kit

Catalog No. 501125078
Change view
Click to view available options
Quantity:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125078 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide
Catalog No. 501125078 Supplier Invitrogen™ Supplier No. 8802684974
Only null left
Contains Sodium azide

Includes

  • MagniSort™ Anti-Mouse CD45.2 Biotin : 200 tests, 20μL/test
  • MagniSort™ Positive Selection Beads A

Designed for the magnetic separation of CD45.2+ cells by positive selection. It has been optimized for the isolation of CD45.2+ cells from mouse spleens or lymph nodes utilizing a biotinylated Anti-Mouse CD45.2 antibody and streptavidin-coated magnetic beads.

  • CD45.2+ cells are bound by antibody and then magnetic beads
  • When placed in a magnetic field, the undesired cells can be separated from CD45.2+ cells by decanting
  • After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD45.2, clone 104
  • When the starting frequency of CD45.2 cells is greater than 75% of the total cells, the amount of MagniSort™ Magnetic Positive Selection Beads to be used can be increased to 40 μL/test to maximize recovery

Specifications

Cell Type CD45.2+
Product Type Positive Selection Kit
No. of Tests 200 tests
Quantity 200 tests
Target Species Mouse

For Research Use Only. Not for use in diagnostic procedures.