missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagniSort™ Mouse CD4 T cell Enrichment Kit

Catalog No. 50728304
Change view
Click to view available options
Quantity:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50728304 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide
Catalog No. 50728304 Supplier Invitrogen™ Supplier No. 8804682174

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Contains Sodium azide

Includes

  • MagniSort™ Mouse CD4 T cell Enrichment Antibody Cocktail : 200 tests, 20μL/test
  • MagniSort™ Negative Selection Beads B

Designed for the magnetic separation of mouse CD4 T cells by negative selection. It has been optimized for the isolation of mouse CD4 T cells from spleens or lymph nodes utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads.

Undesired cells are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave CD4 T cells untouched and free in solution.

The MagniSort™ Mouse CD4 T cell Enrichment Antibody Cocktail contains the following antibodies:
Anti-Mouse CD8 Biotin
Anti-Mouse CD11b Biotin
Anti-Mouse CD19 Biotin
Anti-Mouse CD24 Biotin
Anti-Mouse CD45R (B220) Biotin
Anti-Mouse CD49b (Integrin alpha 2) Biotin
Anti-Mouse Ly-6G (Gr-1) Biotin
Anti-Mouse γδ TCR Biotin
Anti-Mouse TER-119 Biotin

Specifications

Cell Type T Cells (CD4)
Product Type Cell Enrichment Kit
No. of Tests 200 tests
Quantity 200 tests
Target Species Mouse

For Research Use Only. Not for use in diagnostic procedures.