missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagniSort™ Mouse CD4 Positive Selection Kit

Catalog No. 501125071
Change view
Click to view available options
Quantity:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125071 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide
Catalog No. 501125071 Supplier Invitrogen™ Supplier No. 8802684174
Contains Sodium azide

Includes

  • MagniSort Anti-Mouse CD4 Biotin : 200 tests, 20μL/test
  • MagniSort Positive Selection Beads A

Designed for the magnetic separation of CD4+ cells by positive selection. It has been optimized for the isolation of CD4+ cells from mouse spleens or lymph nodes utilizing a biotinylated Anti-Mouse CD4 antibody and streptavidin-coated magnetic beads.

  • CD4+ cells are bound by antibody and then magnetic beads
  • When placed in a magnetic field, the undesired cells can be separated from CD4+ cells by decanting
  • After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD4, clones RM4-5 or GK1.5

Specifications

Cell Type CD4+
Product Type Positive Selection Kit
No. of Tests 200 tests
Quantity 200 tests
Target Species Mouse

For Research Use Only. Not for use in diagnostic procedures.