missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ MagniSort™ Mouse CD335 (NKp46) Positive Selection Kit

Catalog No. 501125062
Change view
Click to view available options
Quantity:
200 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125062 200 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide
Catalog No. 501125062 Supplier Invitrogen™ Supplier No. 8802681774
Contains Sodium azide

Includes

  • MagniSort Anti-Mouse CD335 (NKp46) Biotin : 200 tests, 20μL/test
  • MagniSort Positive Selection Beads A

Designed for the magnetic separation of CD335+ cells by positive selection. It has been optimized for the isolation of CD335+ cells from mouse spleens or lymph nodes utilizing a biotinylated Anti-Mouse CD335+ antibody and streptavidin-coated magnetic beads.

  • CD335+ cells are bound by antibody and then magnetic beads
  • When placed in a magnetic field, the undesired cells can be separated from CD335+ cells by decanting
  • After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD49b (Integrin alpha 2), clone DX5, or Anti-Mouse NK1.1, clone PK136, in C57BL, FVB/N and NZB mice

Specifications

Content And Storage 2° to 8°C. Do not freeze.
For Use With (Application) Magnetic Cell Separation
Format Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Quantity 200 Tests