missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagniSort™ Mouse CD11c Positive Selection Kit

Catalog No. 501125080
Change view
Click to view available options
Quantity:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125080 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide
Catalog No. 501125080 Supplier Invitrogen™ Supplier No. 8802686174
Only null left
Contains Sodium azide

Includes

  • MagniSort™ Mouse CD11c Biotin : 200 tests, 20μL/test
  • MagniSort™ Positive Selection Beads B

Designed for the magnetic separation of CD11c+ cells by positive selection. It has been optimized for the isolation of CD11c+ cells from mouse spleens utilizing a biotinylated Anti-Mouse CD11c antibody and streptavidin-coated magnetic beads.

  • CD11c+ cells are bound by antibody and then magnetic beads
  • When placed in a magnetic field, the undesired cells can be separated from CD11c+ cells by decanting
  • After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD11c, clone N418 and Anti-Mouse CD11b, clone M1/70
  • To reduce non-specific binding, the MagniSort™ Anti-Mouse CD11c Biotin contains Fc Receptor block

Specifications

Cell Type CD11c+
Product Type Positive Selection Kit
No. of Tests 200 tests
Quantity 200 tests
Target Species Mouse

For Research Use Only. Not for use in diagnostic procedures.