missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagniSort™ Mouse CD11b Positive Selection Kit

Catalog No. 501125079
Change view
Click to view available options
Quantity:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125079 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide
Catalog No. 501125079 Supplier Invitrogen™ Supplier No. 8802686074
Contains Sodium azide

Includes

  • MagniSort Mouse CD11b Biotin : 200 tests, 20μL/test
  • MagniSort Positive Selection Beads B

Designed for the magnetic separation of CD11b+ cells by positive selection. It has been optimized for the isolation of CD11b+ cells from mouse bone marrow cells or splenocytes utilizing a biotinylated Anti-Mouse CD11b antibody and streptavidin-coated magnetic beads.

  • CD11b+ cells are bound by antibody and then magnetic beads
  • When placed in a magnetic field, the undesired cells can be separated from CD11b+ cells by decanting
  • After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD11b, clone M1/70

Specifications

Cell Type CD11b+
Product Type Positive Selection Kit
No. of Tests 200 tests
Quantity 200 tests
Target Species Mouse

For Research Use Only. Not for use in diagnostic procedures.