missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagniSort™ Human T cell Enrichment Kit

Catalog No. 50728296
Change view
Click to view available options
Quantity:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50728296 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide
Catalog No. 50728296 Supplier Invitrogen™ Supplier No. 8804681074

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Contains Sodium azide

Includes

  • MagniSort Human T cell Enrichment Antibody Cocktail : 200 tests, 20μL/test
  • MagniSort Negative Selection Beads B

Designed for the magnetic separation of T cells by negative selection. It has been optimized for the isolation of human T cells from normal human peripheral blood mononuclear cells utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads.

Undesired cells are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave T cells untouched and free in solution.

The MagniSort™ Human T cell Enrichment Antibody Cocktail contains the following antibodies:
Anti-Human CD11b Biotin
Anti-Human CD14 Biotin
Anti-Human CD16 Biotin
Anti-Human CD19 Biotin
Anti-Human CD20 Biotin
Anti-Human CD36 Biotin
Anti-Human CD56 Biotin
Anti-Human CD123 Biotin
Anti-Human CD235a Biotin

Specifications

Cell Type T Cells
Product Type Cell Enrichment Kit
No. of Tests 200 tests
Quantity 200 tests
Target Species Human

For Research Use Only. Not for use in diagnostic procedures.