missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ MagniSort™ Human NK cell Enrichment Kit

Catalog No. 501125087
Change view
Click to view available options
Quantity:
200 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125087 200 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide
Catalog No. 501125087 Supplier Invitrogen™ Supplier No. 8804681974
Contains Sodium azide

Includes

  • MagniSort Human NK cell Enrichment Antibody Cocktail : 200 tests, 20μL/test
  • MagniSort Negative Selection Beads B

Designed for the magnetic separation of NK cells by negative selection. It has been optimized for the isolation of human NK cells from normal human peripheral blood mononuclear cells utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads.

Undesired cells, including NKT cells, are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave NK cells untouched and free in solution.

The MagniSort™ Human NK cell Enrichment Antibody Cocktail contains the following antibodies:
Anti-Human CD3 Biotin
Anti-Human CD4 Biotin
Anti-Human CD14 Biotin
Anti-Human CD19 Biotin
Anti-Human CD20 Biotin
Anti-Human CD66b Biotin
Anti-Human CD123 Biotin
Anti-Human CD235a Biotin
Anti-Human HLA-DR Biotin

Specifications

Content And Storage 2° to 8°C. Do not freeze.
For Use With (Application) Magnetic Cell Separation
Format Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Quantity 200 Tests