missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagniSort™ Human Hematopoietic Lineage Depletion Kit

Catalog No. 501128632
Change view
Click to view available options
Quantity:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501128632 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide
Catalog No. 501128632 Supplier Invitrogen™ Supplier No. 8804683674
Contains Sodium azide

Includes

  • MagniSort Human Hematopoietic Lineage Antibody Cocktail : 200 tests, 20μL/test
  • MagniSort Negative Selection Beads B

Designed for the magnetic depletion of lineage positive cells from normal human peripheral blood mononuclear cells. Magnisort Human Hematopoietic Lineage Depletion Kit uses a biotinylated antibody cocktail and streptavidin-coated magnetic beads.

Undesired cells, including T cells, B cells, NK cells, myeloid cells, granulocytes, and red blood cells, are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave lineage negative cells untouched and free in solution.

The MagniSort™ Human Hematopoietic Lineage Antibody Cocktail contains the following antibodies:
Anti-Human CD2 Biotin
Anti-Human CD3 Biotin
Anti-Human CD10 Biotin
Anti-Human CD11b Biotin
Anti-Human CD14 Biotin
Anti-Human CD16 Biotin
Anti-Human CD19 Biotin
Anti-Human CD56 Biotin
Anti-Human CD123 Biotin
Anti-Human CD235a Biotin

Specifications

Cell Type PBMCs
Product Type Cell Depletion Kit
No. of Tests 200 tests
Quantity 200 tests
Target Species Human

For Research Use Only. Not for use in diagnostic procedures.