missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ MagniSort™ Human CD4 T cell 2-Step Enrichment Kit

Catalog No. 501125064
Change view
Click to view available options
Quantity:
200 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125064 200 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide
Catalog No. 501125064 Supplier Invitrogen™ Supplier No. 8802683174
Only null left
Contains Sodium azide

Includes

  • MagniSort Anti-Human CD14 Biotin : 200 tests, 20μL/test
  • MagniSort Anti-Human CD4 Biotin : 200 tests, 20μL/test
  • MagniSort Positive Selection Beads A

Designed for magnetic separation of CD4+ T cells by positive selection after monocyte depletion

    Method
  • Monocytes are bound by Anti-Human CD14 antibody and then magnetic beads
  • When placed in a magnetic field, the monocytes are held in the magnet while the remaining cells are decanted and used for the second step
  • In the second step, the monocyte-depleted cells are bound by Anti-Human CD4 antibody and then magnetic beads
  • Now, when placed in the magnetic field, the CD4+ T cells are held in the magnet while undesired cells can be decanted
  • After enrichment, the purity of selected cells can be verified by staining with Anti-Human CD4, clones OKT4 or SK3

Specifications

Content And Storage 2° to 8°C. Do not freeze.
For Use With (Application) Magnetic Cell Separation
Format Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Quantity 200 Tests