missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ MagniSort™ Human CD4 Memory T cell Enrichment Kit

Catalog No. 50728299
Change view
Click to view available options
Quantity:
200 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50728299 200 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide
Catalog No. 50728299 Supplier Invitrogen™ Supplier No. 8804681374
Contains Sodium azide

Includes

  • MagniSort Human CD4 Memory T cell Enrichment Antibody Cocktail : 200 tests, 20μL/test
  • MagniSort Negative Selection Beads A

Designed for the magnetic separation of CD4 memory T cells by negative selection. It has been optimized for the isolation of human CD4 memory T cells from normal human peripheral blood mononuclear cells utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads.

Undesired cells are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave CD4 memory T cells untouched and free in solution.

The MagniSort™ Human CD4 Memory T cell Enrichment Antibody Cocktail contains the following antibodies:
Anti-Human CD8 Biotin
Anti-Human CD11b Biotin
Anti-Human CD14 Biotin
Anti-Human CD16 Biotin
Anti-Human CD19 Biotin
Anti-Human CD20 Biotin
Anti-Human CD36 Biotin
Anti-Human CD45RA Biotin
Anti-Human CD56 Biotin
Anti-Human CD123 Biotin
Anti-Human CD235a Biotin
Anti-Human γδ TCR Biotin

Specifications

Content And Storage 2° to 8°C. Do not freeze.
For Use With (Application) Magnetic Cell Separation
Format Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Quantity 200 Tests