missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagniSort™ Human CD14 Positive Selection Kit

Catalog No. 501125067
Change view
Click to view available options
Quantity:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125067 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide
Catalog No. 501125067 Supplier Invitrogen™ Supplier No. 8802683474
Contains Sodium azide

Includes

  • MagniSort Anti-Human CD14 Biotin : 200 tests, 20μL/test
  • MagniSort Positive Selection Beads A

Designed for the magnetic separation of CD14+ cells by positive selection. It may also be used for the depletion of CD14+ cells. It has been optimized for use with human peripheral blood monocuclear cells, utilizing a biotinylated Anti-Human CD14 antibody and streptavidin-coated magnetic beads.

  • CD14+ cells are bound by antibody and then magnetic beads
  • When placed in a magnetic field, the CD14- cells can be separated from CD14+ cells by decanting
  • After selection, the purity of selected cells can be verified by staining with Anti-Human CD4, clone RPA-T4 and Anti-Human CD14, clone 61D3

Specifications

Cell Type CD14+
Product Type Positive Selection Kit
No. of Tests 200 tests
Quantity 200 tests
Target Species Human

For Research Use Only. Not for use in diagnostic procedures.