missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagniSort™ Human CD117 (c-Kit) Positive Selection Kit

Catalog No. 501125084
Change view
Click to view available options
Quantity:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125084 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide
Catalog No. 501125084 Supplier Invitrogen™ Supplier No. 8802687074
Contains Sodium azide

Includes

  • MagniSort Anti-Human CD117 (c-Kit) Biotin : 200 tests, 20μL/test
  • MagniSort Positive Selection Beads B

Designed for the magnetic separation of CD117+ cells by positive selection. It has been optimized for the isolation of CD117+ cells from normal human peripheral blood cells.

  • CD117+ cells are bound by antibody and then magnetic beads
  • When placed in a magnetic field, the undesired cells can be separated from CD117+ cells by decanting
  • After positive selection, the purity of selected cells can be verified by staining with Anti-Human CD117, clone YB5.B8, and Anti-Human CD34

Specifications

Cell Type CD117+
Product Type Positive Selection Kit
No. of Tests 200 tests
Quantity 200 tests
Target Species Human

For Research Use Only. Not for use in diagnostic procedures.