missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ MagniSort™ Human B Cell Enrichment Kit, 2nd Generation

Catalog No. 501125093
Change view
Click to view available options
Quantity:
200 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125093 200 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide
Catalog No. 501125093 Supplier Invitrogen™ Supplier No. 8804686774
Only null left
Contains Sodium azide

Includes

  • MagniSort Human B cell Enrichment Antibody Cocktail (2nd Generation) : 200 tests, 20μL/test
  • MagniSort Negative Selection Beads B

Designed for the magnetic separation of mouse B cells by negative selection with improved purity. It has been optimized for the isolation of human B cells from normal human peripheral blood mononuclear cells utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads.

Undesired cells are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave B cells untouched and free in solution.

The MagniSort™ Mouse B cell Enrichment Antibody Cocktail contains the following antibodies:
Anti-Human CD2 Biotin
Anti-Human CD3 Biotin
Anti-Human CD11b Biotin
Anti-Human CD14 Biotin
Anti-Human CD16 Biotin
Anti-Human CD36 Biotin
Anti-Human CD56 Biotin
Anti-Human CD235a Biotin

Specifications

Content And Storage 2° to 8°C. Do not freeze.
For Use With (Application) Magnetic Cell Separation
Format Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Quantity 200 Tests