Learn More
Eisco™ Magnifier with Folding Aluminium Case
Catalog No. S02908
Change view
Click to view available options
Magnification:
10x
Catalog No. Magnification
S02908 10x
1 options
Catalog No. S02908 Supplier Eisco™ Supplier No. PH0502A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Eisco Labs gowland type plano-convex lens made of extra white glass. Features an unbreakable plastic lens holder and an anodized aluminum cover. Ideal for precise optical applications.
  • Unbreakable plastic lens holder and aluminum anodized cover
  • Gowland type, Plano-convex lens of extra white glass
Specifications
Diameter (Englis) Lens 1.5 in.
Diameter (Metric) Lens 38 mm
Product Type Magnifier, Folding
Material Glass
Magnification 10x