Learn More
CHECKERS™ Magnetic Mount Base for Lighted Whips
Catalog No. 19085962
Change view
Click to view available options
Material:
Metal
Catalog No. Material
19085962 Metal
1 options
Catalog No. 19085962 Supplier CHECKERS™ Supplier No. FS7009PCS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Magnetic mount base with handle and 7 foot coiled power cord with lighter plug is recommended for temporary use only, and for whips that are 3 or 5 foot in length
Specifications
Includes 7 ft. coiled power cord with lighter plug
Material Metal
Packaging Quantity 1 Ea.
For Use With (Equipment) Lighted whips