Learn More
Fine Science Tools MAGNETIC FIXATOR RETRACTION SYSTEM
Catalog No. NC1628183
Encompass
Product: Magnetic Fixator Retraction System - Kit for Animals 200g to 5kg/ Item: Complete Retraction System/ Dimensions: 35cm Long/ Maximum Spread: Adjustable/ Alloy: Stainless Steel/ Unit: Full Kit
Catalog No. NC1628183 Supplier Fine Science Tools Supplier No. FST1820050
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Product: Magnetic Fixator Retraction System - Kit for Animals 200g to 5kg/ Item: Complete Retraction System/ Dimensions: 35cm Long/ Maximum Spread: Adjustable/ Alloy: Stainless Steel/ Unit: Full Kit