missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Magnet Stirrup

Catalog No. S15850
Change view
Click to view available options
Grade:
5+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S15850 5+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15850 Supplier United Scientific™ Supplier No. MGSTR01

Simple support for magnets or friction rods.

  • Brass support bar magnets or friction rods. Allows magnets or friction rods to hang freely and turn in a horizontal plane.
  • Hooks are 89 mm apart
  • Dimensions: 3.5 x 2 x 1 in.

Specifications

For Use With (Application) Magnetic Demonstrations
Grade 5+
Product Type Magnet Stirrup

Warning: Cancer and reproductive harm