missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magneson, TCI America™
$32.28 - $62.66
Chemical Identifiers
| CAS | 74-39-5 |
|---|---|
| Molecular Formula | C12H9N3O4 |
| Molecular Weight (g/mol) | 259.22 |
| MDL Number | MFCD00007310 |
| InChI Key | BIFFKKLAAIGZBV-UHFFFAOYSA-N |
| Synonym | Azo Violet, 4-(4-Nitrophenylazo)resorcinol |
| PubChem CID | 5717413 |
| IUPAC Name | 3-hydroxy-4-[2-(4-nitrophenyl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one |
| SMILES | OC1=CC(=O)C=CC1=NNC1=CC=C(C=C1)[N+]([O-])=O |
Chemical Identifiers
| 74-39-5 | |
| 259.22 | |
| BIFFKKLAAIGZBV-UHFFFAOYSA-N | |
| 5717413 | |
| OC1=CC(=O)C=CC1=NNC1=CC=C(C=C1)[N+]([O-])=O |
| C12H9N3O4 | |
| MFCD00007310 | |
| Azo Violet, 4-(4-Nitrophenylazo)resorcinol | |
| 3-hydroxy-4-[2-(4-nitrophenyl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one |
Specifications
| 192°C | |
| C12H9N3O4 | |
| Azo Violet, 4-(4-Nitrophenylazo)resorcinol | |
| OC1=CC(=O)C=CC1=NNC1=CC=C(C=C1)[N+]([O-])=O | |
| 259.22 | |
| 259.22 | |
| Crystal/Powder |
| 74-39-5 | |
| MFCD00007310 | |
| BIFFKKLAAIGZBV-UHFFFAOYSA-N | |
| 3-hydroxy-4-[2-(4-nitrophenyl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one | |
| 5717413 | |
| 1 g | |
| Magneson |
Safety and Handling
RTECSNumber : VH2810000
TSCA : Yes