missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magneson, TCI America™
Specifications
Specifications
| Melting Point | 192°C |
| Quantity | 25 g |
| Physical Form | Crystal/Powder |
| Formula Weight | 259.22 |
| Chemical Name or Material | Magneson |
Safety and Handling
| RTECSNumber | VH2810000 |
| TSCA | Yes |