missing translation for 'onlineSavingsMsg'
Learn More

Magneson, TCI America™

Catalog No. N01351G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N01351G 1 g
N013525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N01351G Supplier TCI America Supplier No. N01351G

Chemical Identifiers

CAS 74-39-5
Molecular Formula C12H9N3O4
Molecular Weight (g/mol) 259.22
MDL Number MFCD00007310
InChI Key BIFFKKLAAIGZBV-UHFFFAOYSA-N
Synonym Azo Violet, 4-(4-Nitrophenylazo)resorcinol
PubChem CID 5717413
IUPAC Name 3-hydroxy-4-[2-(4-nitrophenyl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one
SMILES OC1=CC(=O)C=CC1=NNC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 192°C
Formula Weight 259.22
Quantity 1 g
Physical Form Crystal/Powder
Chemical Name or Material Magneson
RTECSNumber VH2810000
TSCA Yes