missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium turnings, 99.8% (metals basis), Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Silver |
| Appearance | Silver-colored turnings |
| Density | 1.738g/cm3 at 20°C |
| Quantity | 500 g |
| Odor | Odorless |
| Vapor Pressure | 1.33 hPa (1mm Hg) at 621°C |
| Assay Percent Range | 99.8% (Metals basis) |
| Formula Weight | 24.3 |
| Chemical Name or Material | Magnesium |
Safety and Handling
| Hazard Category | H228-H252-H261 |
| Precautionary Statement | P210-P223-P231+P232-P235+P410-P240-P241-P280-P335+P334-P370+P378s-P501c |
| DOTInformation | DOT Class: 4.1, Packaging Group: III |
| EINECSNumber | 231-104-6 |
| RTECSNumber | OM2100000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only