missing translation for 'onlineSavingsMsg'
Learn More

Magnesium turnings, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog No. AA10232A4
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA10232A4 2.5 kg
AA1023236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA10232A4 Supplier Thermo Scientific Chemicals Supplier No. 010232A4

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Form: 3.2mm (0.13 in.) wide

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-95-4
Molecular Formula Mg
Molecular Weight (g/mol) 24.30
MDL Number MFCD00085308
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
Synonym magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
PubChem CID 5462224
ChEBI CHEBI:25107
IUPAC Name magnesium
SMILES [Mg]

Specifications

Color Silver
Appearance Silver-colored turnings
Quantity 2.5 kg
Odor Odorless
Vapor Pressure 1.33 hPa (1mm Hg) at 621°C
Assay Percent Range 99.8% (Metals basis)
Formula Weight 24.3
Chemical Name or Material Magnesium
Hazard Category H228-H252-H261
Precautionary Statement P210-P223-P231+P232-P235+P410-P240-P241-P280-P335+P334-P370+P378s-P501c
DOTInformation DOT Class: 4.1, Packaging Group: III
EINECSNumber 231-104-6
RTECSNumber OM2100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only