missing translation for 'onlineSavingsMsg'
Learn More

Magnesium turnings, 1cm (0.4in) & down, Puratronic™, 99.98% (metals basis), Thermo Scientific Chemicals

Catalog No. p-7048469
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1063006 5 g
AA1063022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1063006 Supplier Thermo Scientific Chemicals Supplier No. 01063006
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-95-4
Molecular Formula Mg
Molecular Weight (g/mol) 24.30
MDL Number MFCD00085308
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
Synonym magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
PubChem CID 5462224
ChEBI CHEBI:25107
IUPAC Name magnesium
SMILES [Mg]

Specifications

Quantity 5 g
Odor Odorless
Particle Size 1cm (0.4 in.) & down
UN Number UN1869
Percent Purity 99.98%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Magnesium turnings
Hazard Category H228-H252-H261
Hazard Statement GHS H Statement
H228-H251-H261
Flammable solid.
Self-heating: may catch fire.
In contact with water releases flammable gas.
Precautionary Statement P210-P223-P231+P232-P235+P410-P240-P241-P280-P335+P334-P370+P378s-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: MAGNESIUM
EINECSNumber 231-104-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only