missing translation for 'onlineSavingsMsg'
Learn More

Magnesium titanium oxide, 99% (metals basis)

Catalog No. AA11398A1
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA11398A1 1 kg
AA1139830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA11398A1 Supplier Thermo Scientific Chemicals Supplier No. 011398A1
Only null left

CAS: 12032-30-3 | MgO3Ti | 120.169 g/mol

Magnesium titanium oxide is used in the primary chemical intermediates and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12032-30-3
Molecular Formula MgO3Ti
Molecular Weight (g/mol) 120.169
MDL Number MFCD00016198
InChI Key XSETZKVZGUWPFM-UHFFFAOYSA-N
Synonym magnesium titanate, magnesium dititanate, karrooite, ticon 75, magnesium titanium trioxide, atd 16, magnesium titanate iv mgtio3, titanium magnesium oxide timgo3, magnesium dititanium pentoxide, magnesium titanate mgti2o5
PubChem CID 159407
IUPAC Name magnesium;oxygen(2-);titanium(4+)
SMILES [O-2].[O-2].[O-2].[Mg+2].[Ti+4]

Specifications

Melting Point 1610°C
Color White
Physical Form Powder
Quantity 1 kg
Solubility Information Insoluble in water.
Formula Weight 120.21
Percent Purity 99%
Odor Odorless
Density 3.36 g/mL
Assay (metals basis)
Chemical Name or Material Magnesium titanium oxide
Show More Show Less
EINECSNumber 234-766-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only