missing translation for 'onlineSavingsMsg'
Learn More

Magnesium thiosulfate hexahydrate, 99%

Catalog No. p-7080033
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4611422 100 g
AA4611436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4611422 Supplier Thermo Scientific Chemicals Supplier No. 04611422
Only null left

CAS: 13446-30-5 | H12MgO9S2 | 244.512 g/mol

Magnesium hyposulfite finds application in the treatment of certain skin diseases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13446-30-5
Molecular Formula H12MgO9S2
Molecular Weight (g/mol) 244.512
MDL Number MFCD00169871
InChI Key CQDMJJVHDPDPHO-UHFFFAOYSA-L
Synonym magnesium thiosulfate hexahydrate, unii-59481j5eha, thiosulfuric acid h2s2o3 , magnesium salt 1:1 , hexahydrate 9ci, magnesium 2+ hypo hexahydrate, magnesium thiosulfate hexahydrate 100g, magnesium 2+ ion hexahydrate thiosulphate
PubChem CID 26002
IUPAC Name magnesium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;hexahydrate
SMILES O.O.O.O.O.O.[O-]S(=O)(=S)[O-].[Mg+2]

Specifications

Melting Point 170°C (in -3H2O)
Color White
Physical Form Crystalline
Quantity 100 g
Linear Formula MgS2O3·6H2O
Merck Index 14,5694
Solubility Information Soluble in water.
Formula Weight 244.53 (136.44 Anhydrous)
Percent Purity 99%
Density 1.818 g/mL
Chemical Name or Material Magnesium thiosulfate hexahydrate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-340-5
RTECSNumber OM4610000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only