missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium sulfate hydrate, Puriss., meets analytical spec. of DAC, dried, 99.0-101.0% MgSO4 basis (in dried substance), Honeywell Fluka™
Puriss., meets analytical specification of DAC, dried, 99.0-101.0% MgSO4 basis (in dried substance)
Supplier: Honeywell-Fluka 131436X1KG
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Magnesium Sulfate Hydrate | |
| 22189-08-8 | |
| MgO4S | |
| MFCD00149787 | |
| magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot | |
| [Mg++].[O-]S([O-])(=O)=O | |
| 120.36 | |
| CHEBI:32599 | |
| 99.0 to 101.0% (MgSO4 basis (in dried substance)) |
| Meets analytical specification of DAC | |
| 6 x 1 kg | |
| MgSO4 · aq | |
| NONH for all modes of transport | |
| CSNNHWWHGAXBCP-UHFFFAOYSA-L | |
| magnesium;sulfate | |
| 24083 | |
| 120.37 (anhydrous basis) | |
| puriss. |
Chemical Identifiers
| 22189-08-8 | |
| 120.36 | |
| CSNNHWWHGAXBCP-UHFFFAOYSA-L | |
| 24083 | |
| [Mg++].[O-]S([O-])(=O)=O |
| MgO4S | |
| MFCD00149787 | |
| magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot | |
| CHEBI:32599 |
Safety and Handling
EINECSNumber : 231-298-2