missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate Heptahydrate (USP/FCC/EP), Fisher Chemical™

Catalog No. p-32408
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M80500 500 g Poly Bottle
M80212 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M80500 Supplier Thermo Fisher Scientific Supplier No. M80500
Only null left

Chemical Identifiers

CAS 10034-99-8
Molecular Formula H14MgO11S
Molecular Weight (g/mol) 246.47
MDL Number MFCD00149785
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
Synonym magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
PubChem CID 24843
ChEBI CHEBI:31795
IUPAC Name magnesium(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

Specifications

Melting Point 770°C
Color White
pH 5 to 8.2
Physical Form Solid
Quantity 500 g
Linear Formula MgSO4·7H2O
Formula Weight 246.48
Percent Purity 99.0 to 100.5% (USP)
Grade EP/FCC/USP
Identification Pass Test (EP), Pass Test (EP/FCC/USP), Pass Test (FCC), Pass Test (USP)
Loss on Drying 48.0 to 52.0% (EP)
Loss on Ignition 40.0 to 52.0% (FCC), 40.0 to 52.0% (FCC/USP), 40.0 to 52.0% (USP)
Packaging Poly Bottle
Vapor Pressure <0.1 mmHg at 20°C
Density 1.678g/cm³
Chemical Name or Material Magnesium Sulfate Heptahydrate
Show More Show Less