missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium sulfate heptahydrate, puriss. p.a., ACS reagent, Honeywell Fluka™
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 500 g |
| Linear Formula | MgSO4 · 7H2O |
| UN Number | NONH for all modes of transport |
| Formula Weight | 246.47g/mol |
| Percent Purity | ≥99% (KT) |
| Grade | ACS/puriss. p.a. |
| Vapor Pressure | 0.1 mmHg (20°C) |
| Density | Vapor Density: <0.01 (vs air) |
| Chemical Name or Material | Magnesium sulfate heptahydrate |
Safety and Handling
| RTECSNumber | OM4508000 |