missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate Heptahydrate, Purified (Epsom Salts), Reagents

Catalog No. 01337186
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337186 12 kg
01337185 2.5 kg
01337184 500 g
01337183 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01337186 Supplier Labchem Inc Supplier No. C224000012KG
Only null left

Magnesium Sulfate Heptahydrate, Purified (Epsom Salts), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Magnesium Sulfate Heptahydrate, Purified (Epsom Salts)
Boiling Point 200°C
Color Colorless/White
CAS 10034-99-8
Physical Form Solid
Quantity 12 kg
Molecular Formula MgSO4, 7H2O
Molecular Weight (g/mol) 246.4746
Grade Pure
Packaging Poly Pail
Specific Gravity 1.67
Show More Show Less