missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium sulfate heptahydrate, meets analytical spec. of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance), Honeywell Fluka™
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 6 x 1 kg |
| Linear Formula | MgSO4 · 7H2O |
| UN Number | NONH for all modes of transport |
| Formula Weight | 246.47 |
| Percent Purity | 99.5 to 100.5% (calc. to the dried substance) |
| Vapor Pressure | 0.1mmHg (20°C) |
| Density | Vapor Density: <0.01 (vs air) |
| Chemical Name or Material | Magnesium Sulfate Heptahydrate, Meets analytical specification of Ph. Eur.BP, USP, FCC |
Safety and Handling
| EINECSNumber | 231-298-2 |
| RTECSNumber | OM4508000 |