missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium sulfate heptahydrate, meets analytical spec. of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance), Honeywell Fluka™
Meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance)
Supplier: Honeywell-Fluka 1314225KG
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Magnesium Sulfate Heptahydrate | |
| 10034-99-8 | |
| H14MgO11S | |
| MFCD00149785 | |
| magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate | |
| O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O | |
| 246.47 | |
| CHEBI:31795 | |
| 99.5 to 100.5% (calc. to the dried substance) | |
| 0.1mmHg (20°C) |
| Meets analytical specification of Ph. Eur.BP, USP, FCC | |
| 2.5 kg | |
| MgSO4 · 7H2O | |
| NONH for all modes of transport | |
| WRUGWIBCXHJTDG-UHFFFAOYSA-L | |
| magnesium;sulfate;heptahydrate | |
| 24843 | |
| 246.47g/mol | |
| Plastic Bottle | |
| Vapor Density: <0.01 (vs air) |
Chemical Identifiers
| 10034-99-8 | |
| 246.47 | |
| WRUGWIBCXHJTDG-UHFFFAOYSA-L | |
| 24843 | |
| magnesium;sulfate;heptahydrate |
| H14MgO11S | |
| MFCD00149785 | |
| magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate | |
| CHEBI:31795 | |
| O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O |
Safety and Handling
EINECSNumber : 231-298-2
RTECSNumber : OM4508000