missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfate heptahydrate, meets analytical spec. of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance), Honeywell Fluka™

Catalog No. 6001968
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
4 x 5 kg
6 x 1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001968 1 kg
6001969 2.5 kg
6001970 5 kg
60044774 6 x 1 kg
60044773 4 x 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 6001968 Supplier Honeywell Chemicals Supplier No. 131421KG
Only null left

Meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 10034-99-8
Molecular Formula H14MgO11S
Molecular Weight (g/mol) 246.47
MDL Number MFCD00149785
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
Synonym magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
PubChem CID 24843
ChEBI CHEBI:31795
IUPAC Name magnesium;sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

Specifications

Quantity 1 kg
Linear Formula MgSO4 · 7H2O
UN Number NONH for all modes of transport
Formula Weight 246.47g/mol
Percent Purity 99.5 to 100.5% (calc. to the dried substance)
Packaging Plastic Bottle
Vapor Pressure 0.1mmHg (20°C)
Density Vapor Density: <0.01 (vs air)
Chemical Name or Material Magnesium Sulfate Heptahydrate, Meets analytical specification of Ph. Eur.BP, USP, FCC
EINECSNumber 231-298-2
RTECSNumber OM4508000