missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate, Heptahydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical

Catalog No. 18611464
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611464 50 kg Poly Bottle
18611459 125 g Poly Bottle
18611460 500 g Poly Bottle
18611461 2.5 kg Poly Bottle
18611462 12 kg Poly Bottle
18611463 25 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18611464 Supplier Spectrum Chemical Mfg Cor Supplier No. M106550KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10034-99-8
Molecular Formula H14MgO11S
Molecular Weight (g/mol) 246.47
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

Specifications

pH 5.0 to 8.2
Physical Form Crystalline Powder, Needles or Crystals
CAS Max % 1
Quantity 50 kg
Packaging Poly Bottle
Percent Purity 98 to 102%
Grade ACS/Reagent
Chemical Name or Material Magnesium Sulfate, Heptahydrate, Crystal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.