missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfate heptahydrate, Cell Culture Reagent

Catalog No. p-7042244
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6183936 500 g
AAJ61839A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6183936 Supplier Thermo Scientific Chemicals Supplier No. J6183936
Only null left

CAS: 10034-99-8 | H14MgO11S | 246.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10034-99-8
Molecular Formula H14MgO11S
Molecular Weight (g/mol) 246.47
MDL Number MFCD00149785
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
Synonym magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
PubChem CID 24843
ChEBI CHEBI:31795
IUPAC Name magnesium;sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

Specifications

Color White
Physical Form Powder
Quantity 500 g
Linear Formula MgSO4·7H2O
Merck Index 14,5691
Formula Weight 246.48 (120.37 Anhydrous)
Grade Cell Culture Reagent
Odor Odorless
Density 1.68
Chemical Name or Material Magnesium sulfate heptahydrate
EINECSNumber 231-298-2
RTECSNumber OM4508000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only