missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Sulfate, Heptahydrate, ACS Grade, 98.0 to 102.0 %, LabChem™
Specifications
Specifications
| Color | White |
| CAS Max % | 100 |
| Physical Form | Crystals |
| Quantity | 500 g |
| Linear Formula | MgSO4·7H2O |
| Solubility Information | Soluble in water |
| Formula Weight | 246.47 |
| Percent Purity | 98.0 to 102.0% |
| Grade | ACS |
| Identification | Passes Test |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement Solution is not hazardous. |
| Precautionary Statement | GHS P Statement If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes. If swallowed, get medical attention. |
| Recommended Storage | Room Temperature |