missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfate heptahydrate, ACS, 98.0-102.0%

Catalog No. AA1159622
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1159622 100 g
AA1159636 500 g
AA11596A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1159622 Supplier Thermo Scientific Chemicals Supplier No. 01159622

For Fireproofing, Textiles, Catalyst Carrier | CAS: 10034-99-8 | H14MgO11S | 246.47 g/mol

Magnesium sulfate produced by appropriate process finds use in several pharmaceutical preparations of magnesium, usually known as epsom salt. It is used as a micronutrient in agriculture, brewing salt in production of certain alcoholic products, electrolyte to prepare metal salts, coagulant, and ingredient in bath salts for use in floatation therapy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10034-99-8
Molecular Formula H14MgO11S
Molecular Weight (g/mol) 246.47
MDL Number MFCD00149785
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
Synonym magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
PubChem CID 24843
ChEBI CHEBI:31795
IUPAC Name magnesium;sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

Specifications

Color White
Physical Form Crystals
Quantity 100 g
Assay Percent Range 98.0-102.0%
Solubility Information Soluble in in water and acid. Slightly soluble in alcohol and glycerol.
Formula Weight 246.48 (120.37 Anhydrous)
Grade ACS
Odor Odorless
Appearance White crystals
Density 1.68 g/cm3 at 20°C
Chemical Name or Material Magnesium sulfate heptahydrate, heptahydrate
Show More Show Less
EINECSNumber 231-298-2
RTECSNumber OM4508000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only