missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfate heptahydrate, 99+%

Catalog No. p-7021946
Change view
Click to view available options
Quantity:
1000 g
5000 g
Catalog No. Quantity
AAA144910B 1000 g
AAA144910I 5000 g
2 options

Catalog No. AAA144910B

Supplier: Thermo Scientific Chemicals A144910B

Only null left

CAS: 10034-99-8 | H14MgO11S | 246.47 g/mol

Magnesium sulfate, produced by appropriate processes, finds use in several pharmaceutical preparations of magnesium usually known as epsom salt. It is used as a micronutrient in agriculture, for brewing salt in the production of certain alcoholic products, as an electrolyte to prepare metal salts, as a coagulant and as an ingredient in bath salts for use in floatation therapy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium sulfate, produced by appropriate processes, finds use in several pharmaceutical preparations of magnesium usually known as epsom salt. It is used as a micronutrient in agriculture, for brewing salt in the production of certain alcoholic products, as an electrolyte to prepare metal salts, as a coagulant and as an ingredient in bath salts for use in floatation therapy.

Solubility
Soluble in in water and acid. Slightly soluble in alcohol and glycerol.

Notes
Incompatible with strong oxidizers.

Chemical Identifiers

CAS 10034-99-8
Molecular Formula H14MgO11S
Molecular Weight (g/mol) 246.47
MDL Number MFCD00149785
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
Synonym magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
PubChem CID 24843
ChEBI CHEBI:31795
IUPAC Name magnesium;sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

Specifications

Quantity 1000 g
Merck Index 14,5691
Solubility Information Soluble in in water and acid. Slightly soluble in alcohol and glycerol.
Formula Weight 246.48 (120.37 Anhydrous)
Percent Purity ≥99%
Odor Odorless
Density 1.68
Chemical Name or Material Magnesium sulfate heptahydrate
EINECSNumber 231-298-2
RTECSNumber OM4508000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only