missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate Heptahydrate, ≥99%, MP Biomedicals

$99.57 - $131.83

Chemical Identifiers

CAS 10034-99-8
Molecular Formula H14MgO11S
Molecular Weight (g/mol) 246.47
MDL Number MFCD00149785
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
Synonym magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
PubChem CID 24843
ChEBI CHEBI:31795
IUPAC Name magnesium(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19483390
View Documents
MP Biomedicals Inc
0219483390
500 g
Each for $99.57
Only null left
 
ICN-19483391
View Documents
MP Biomedicals Inc
0219483391
1 kg
Each for $131.83
Only null left
 
Description

Description

Specifications

Chemical Identifiers

10034-99-8
246.47
WRUGWIBCXHJTDG-UHFFFAOYSA-L
24843
magnesium(2+) heptahydrate sulfate
H14MgO11S
MFCD00149785
magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
CHEBI:31795
O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

Specifications

10034-99-8
≥99%
MFCD00149785
WRUGWIBCXHJTDG-UHFFFAOYSA-L
magnesium(2+) heptahydrate sulfate
24843
246.5
500 g
H14MgO11S
magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
246.47
CHEBI:31795
Magnesium Sulfate Heptahydrate
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
missing translation for 'documents'

missing translation for 'documents'