missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfate heptahydrate, 99%, for biochemistry, Thermo Scientific Chemicals

$208.56 - $208.56

Chemical Identifiers

CAS 10034-99-8
Molecular Formula H14MgO11S
Molecular Weight (g/mol) 246.47
MDL Number MFCD00149785
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
Synonym magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
PubChem CID 24843
ChEBI CHEBI:31795
IUPAC Name magnesium;sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC447165000
SDS View Documents
Thermo Scientific Chemicals
447165000
500 g Plastic bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10034-99-8
246.47
WRUGWIBCXHJTDG-UHFFFAOYSA-L
24843
magnesium;sulfate;heptahydrate
H14MgO11S
MFCD00149785
magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
CHEBI:31795
O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

Specifications

770.0°C
500 g
H14MgO11S
MFCD00149785
magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
WRUGWIBCXHJTDG-UHFFFAOYSA-L
magnesium;sulfate;heptahydrate
24843
246.48
Biochemical
Magnesium sulfate heptahydrate
10034-99-8
98.5% min. (Complexometry)
MgSO4·7H2O
15,5755
(1 M in water) Clear colorless
O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
246.47
CHEBI:31795
99%
Plastic bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only