missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfate heptahydrate, 98+%, ACS reagent

CAS: 10034-99-8 | H14MgO11S | 246.47 g/mol

Supplier:  Thermo Scientific Chemicals 423900050

 View more versions of this product

Catalog No. AC423900050


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Magnesium sulfate heptahydrate
10034-99-8
5.0 to 8.2 (5% soln. at 25°C)
5 kg
H14MgO11S
MFCD00149785
magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
WRUGWIBCXHJTDG-UHFFFAOYSA-L
magnesium;sulfate;heptahydrate
24843
246.48
ACS Reagent
770.0°C
White
Crystals
98.0 to 102% (Complexometry)
MgSO4·7H2O
15, 5755
Solubility in water: 710g/L (20°C). Other solubilities: soluble in glycerol, sparingly soluble in alcohol
O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
246.47
CHEBI:31795
≥98%
Plastic Drum

Chemical Identifiers

10034-99-8
246.47
WRUGWIBCXHJTDG-UHFFFAOYSA-L
24843
magnesium;sulfate;heptahydrate
H14MgO11S
MFCD00149785
magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
CHEBI:31795
O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : OM4508000

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only