missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfate, dried, contains ca 1-2 mol water of hydration

Catalog No. AAL1373936
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1373936 500 g
AAL1373922 100 g
AAL137390E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1373936 Supplier Thermo Scientific Chemicals Supplier No. L1373936
Only null left

CAS: 22189-08-8 | MgO4S | 120.36 g/mol

Suitable for the preliminary drying of organic liquids, for example solvent extracts from reaction workups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22189-08-8
Molecular Formula MgO4S
Molecular Weight (g/mol) 120.36
MDL Number MFCD00149787
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
PubChem CID 24083
ChEBI CHEBI:32599
IUPAC Name magnesium;sulfate
SMILES [Mg++].[O-]S([O-])(=O)=O

Specifications

Quantity 500 g
Merck Index 14,5691
Formula Weight 120.37 (Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Magnesium sulfate, contains ≈1 to 2 mol water of hydration
EINECSNumber 231-298-2
RTECSNumber OM4500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only