missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium sulfate (Cell Culture), MP Biomedicals
Cell Culture Reagent
$243.84 - $243.84
Chemical Identifiers
| CAS | 7487-88-9 |
|---|---|
| Molecular Formula | MgO4S |
| Molecular Weight (g/mol) | 120.36 |
| MDL Number | MFCD00011110 |
| InChI Key | CSNNHWWHGAXBCP-UHFFFAOYSA-L |
| Synonym | magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot |
| PubChem CID | 24083 |
| ChEBI | CHEBI:32599 |
| SMILES | [Mg++].[O-]S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15013691
|
MP Biomedicals Inc
0215013691 |
1 kg |
N/A
|
|
|||||
Chemical Identifiers
| 7487-88-9 | |
| 120.36 | |
| CSNNHWWHGAXBCP-UHFFFAOYSA-L | |
| 24083 | |
| [Mg++].[O-]S([O-])(=O)=O |
| MgO4S | |
| MFCD00011110 | |
| magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot | |
| CHEBI:32599 |
Specifications
| 7487-88-9 | |
| MgO4S | |
| magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot | |
| [Mg++].[O-]S([O-])(=O)=O | |
| 120.36 | |
| CHEBI:32599 | |
| Magnesium Sulfate |
| 1 kg | |
| MFCD00011110 | |
| CSNNHWWHGAXBCP-UHFFFAOYSA-L | |
| magnesium(2+) sulfate | |
| 24083 | |
| 120.4 |