missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate, Anhydrous, Powder, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor M106325KG

 View more versions of this product

Catalog No. 18611456


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

7487-88-9
2.5 kg
CSNNHWWHGAXBCP-UHFFFAOYSA-L
magnesium(2+) sulfate
99%
0.05
1
MgO4S
[Mg++].[O-]S([O-])(=O)=O
120.36
Reagent
Poly Bottle

Chemical Identifiers

7487-88-9
120.36
magnesium(2+) sulfate
MgO4S
CSNNHWWHGAXBCP-UHFFFAOYSA-L
[Mg++].[O-]S([O-])(=O)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.