missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate, Anhydrous, Powder, 99%, Spectrum™ Chemical

Catalog No. 18611454
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611454 125 g Poly Bottle
18611455 500 g Poly Bottle
18611456 2.5 kg Poly Bottle
18611457 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18611454 Supplier Spectrum Chemical Mfg Cor Supplier No. M1063125GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 7487-88-9
Molecular Formula MgO4S
Molecular Weight (g/mol) 120.36
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
IUPAC Name magnesium(2+) sulfate
SMILES [Mg++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 99%
Grade Reagent
Loss on Ignition 0.05
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.