missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfate, 99%, extra pure, dried, contains 3 to 4 moles of water

CAS: 22189-08-8 | MgO4S | 120.36 g/mol

$129.10 - $129.10

Chemical Identifiers

CAS 22189-08-8
Molecular Formula MgO4S
Molecular Weight (g/mol) 120.36
MDL Number MFCD00149787
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
PubChem CID 24083
ChEBI CHEBI:32599
SMILES [Mg++].[O-]S([O-])(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC196850010
SDS View Documents
Thermo Scientific Chemicals
196850010
1 kg Plastic Bottle
Each for $129.10
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
Specifications

Chemical Identifiers

22189-08-8
120.36
CSNNHWWHGAXBCP-UHFFFAOYSA-L
24083
[Mg++].[O-]S([O-])(=O)=O
MgO4S
MFCD00149787
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
CHEBI:32599

Specifications

1124.0°C
White
1 kg
MgO4S
MFCD00149787
Solubility in water: 360g/L (20°C). Other solubilities: soluble in glycerol, sparingly soluble in alcohol
[Mg++].[O-]S([O-])(=O)=O
120.36
CHEBI:32599
99%
26% to 32% (400 to 500 °C)
2.66
Magnesium sulfate, Dried, contains 3 to 4 Moles of Water
22189-08-8
Crystalline Powder
98.5% min. (On dried substance) (Complexometry)
MgSO4·xH2O
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
CSNNHWWHGAXBCP-UHFFFAOYSA-L
magnesium(2+) sulfate
24083
120.36
Extra Pure
Plastic Bottle
2.6600g/mL
Videos
Safety and Handling

Safety and Handling

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only