missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium sulfate, 99%, for analysis, anhydrous
CAS: 7487-88-9 | MgO4S | 120.36 g/mol
Supplier: Thermo Scientific Chemicals 446890010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Magnesium sulfate | |
| 7487-88-9 | |
| 98.5% min. (Complexometry) | |
| MgSO4 | |
| 15,5755 | |
| Solubility in water: 26g/100mL (0°C). | |
| [Mg++].[O-]S([O-])(=O)=O | |
| 120.36 | |
| CHEBI:32599 | |
| 99% | |
| Plastic bottle |
| 1124.0°C | |
| 1 kg | |
| MgO4S | |
| MFCD00011110 | |
| magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot | |
| CSNNHWWHGAXBCP-UHFFFAOYSA-L | |
| magnesium;sulfate | |
| 24083 | |
| 120.37 | |
| Analytical |
Chemical Identifiers
| 7487-88-9 | |
| 120.36 | |
| CSNNHWWHGAXBCP-UHFFFAOYSA-L | |
| 24083 | |
| magnesium;sulfate |
| MgO4S | |
| MFCD00011110 | |
| magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot | |
| CHEBI:32599 | |
| [Mg++].[O-]S([O-])(=O)=O |
Safety and Handling
EINECSNumber : 231-298-2
RUO – Research Use Only