missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Sulfate, 7-Hydrate, Crystal, Multi-Compendial, U.S.P., J.T. Baker™
Supplier: Avantor J.T.Baker 250405
Specifications
| Magnesium Sulfate | |
| 10034-99-8 | |
| 2.5 kg | |
| MFCD00149785 | |
| WRUGWIBCXHJTDG-UHFFFAOYSA-L | |
| magnesium;sulfate;heptahydrate | |
| 24843 | |
| 246.47 | |
| Poly Bottle |
| 7-Hydrate | |
| Crystals | |
| H14MgO11S | |
| magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate | |
| O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O | |
| 246.47 | |
| CHEBI:31795 | |
| Multi-Compendial/USP | |
| 1L = 1.67kg |
Chemical Identifiers
| 10034-99-8 | |
| 246.47 | |
| WRUGWIBCXHJTDG-UHFFFAOYSA-L | |
| 24843 | |
| magnesium;sulfate;heptahydrate |
| H14MgO11S | |
| MFCD00149785 | |
| magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate | |
| CHEBI:31795 | |
| O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O |