missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfamate hydrate, 99%

Catalog No. AA3938130
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3938130 250 g
AA3938118 50 g
AA39381A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3938130 Supplier Thermo Scientific Chemicals Supplier No. 03938130

CAS: 13770-91-7 | H4MgN2O6S2 | 216.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13770-91-7
Molecular Formula H4MgN2O6S2
MDL Number MFCD00210632
InChI Key YZVJHCGMTYDKFR-UHFFFAOYSA-L
Synonym magnesium disulfamate, magnesium sulfamate, magnesium disulphamate, sulfamic acid, magnesium salt 2:1, magnesium 2+ ion disulfamate
PubChem CID 166902
IUPAC Name magnesium;disulfamate

Specifications

Quantity 250 g
SMILES NS(=O)(=O)[O-].NS(=O)(=O)[O-].[Mg+2]
Molecular Weight (g/mol) 216.465
Formula Weight 216.48 (Anhydrous)
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Magnesium sulfamate hydrate
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 237-398-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only