missing translation for 'onlineSavingsMsg'
Learn More

Magnesium stearate

Catalog No. AAA1624430
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1624430 250 g
AAA162440B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1624430 Supplier Thermo Scientific Chemicals Supplier No. A1624430
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 557-04-0 | C36H70MgO4 | 591.257 g/mol

Magnesium stearate is involved in the production of medical tablets, capsules, powders and polymer formulation. It is also used as a lubricant for tablets, anti-adherent, in dry coating and as a binding agent. It is an important ingredient in baby formulas. Further, it is used in the hydrogenation process. In addition to this, it is used to bind the sugar in hard candies like mints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble in benzene. Insoluble in water, alcohol and ether.

Notes
Incompatible with strong oxidizing agents, acids and alkalis.

Chemical Identifiers

CAS 557-04-0
Molecular Formula C36H70MgO4
Molecular Weight (g/mol) 591.257
MDL Number MFCD00036391
InChI Key HQKMJHAJHXVSDF-UHFFFAOYSA-L
Synonym magnesium stearate, magnesium octadecanoate, magnesium distearate, synpro 90, octadecanoic acid, magnesium salt, dibasic magnesium stearate, petrac mg 20nf, ns-m salt, sm-p, stearic acid, magnesium salt
PubChem CID 11177
ChEBI CHEBI:9254
IUPAC Name magnesium;octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Mg+2]

Specifications

Melting Point 148°C to 152°C
Quantity 250 g
Solubility Information Slightly soluble in benzene. Insoluble in water,alcohol and ether.
Formula Weight 591.27
Chemical Name or Material Magnesium stearate
EINECSNumber 209-150-3
RTECSNumber WI4390000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only